Assay Details
Gene Symbol: Cct6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014228.1 | NP_001014250.1 | 2 - 3 | 280 | 74 | |
GenBank mRNA | BC079020.1 | - | 2 - 3 | 280 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
