Assay Details
Gene Symbol: Wbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138975.1 | NP_620431.1 | 3 - 4 | 304 | 91 | |
XM_006247705.2 | XP_006247767.1 | 3 - 4 | 384 | 91 | ||
GenBank mRNA | AF499026.1 | - | 3 - 4 | 304 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
