Assay Details
Gene Symbol: Wbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138975.1 | NP_620431.1 | 6 - 7 | 663 | 117 | |
XM_006247705.2 | XP_006247767.1 | 5 - 6 | 608 | 117 | ||
GenBank mRNA | AF499026.1 | - | 6 - 7 | 663 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
