Assay Details
Gene Symbol: Map2k4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030023.1 | NP_001025194.1 | 1 - 2 | 210 | 130 | |
GenBank mRNA | AY880882.1 | - | 1 - 2 | 210 | 130 | |
KC795688.1 | - | 2 - 3 | 243 | 130 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
