Assay Details
Gene Symbol: Scrn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012142.1 | NP_001012142.1 | 1 - 2 | 53 | 74 | |
GenBank mRNA | BC078939.1 | - | 1 - 2 | 53 | 7108372 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC078939.1, NM_001012142.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
