Assay Details
Gene Symbol: Slc16a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006247931.2 | XP_006247993.1 | 2 - 3 | 183 | 118 | |
XM_006247932.2 | XP_006247994.1 | 2 - 3 | 475 | 118 | ||
XM_008768513.1 | XP_008766735.1 | 2 - 3 | 2907 | 118 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006247931.2, XM_006247932.2, XM_008768513.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
