Assay Details
Gene Symbol: Pgap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143895.1 | NP_001137367.1 | 3 - 4 | 474 | 115 | |
GenBank mRNA | BC161914.1 | - | 3 - 4 | 441 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
