Assay Details
Gene Symbol: Sar1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009622.1 | NP_001009622.1 | 1 - 2 | 67 | 89 | |
GenBank mRNA | BC088842.1 | - | 1 - 2 | 67 | 7303357 | 89 |
FQ217751.1 | - | 1 - 2 | 128 | 89 | ||
FQ219381.1 | - | 1 - 2 | 128 | 89 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC088842.1,NM_001009622.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
