Assay Details
Gene Symbol: Sar1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009622.1 | NP_001009622.1 | 3 - 4 | 270 | 63 | |
GenBank mRNA | BC088842.1 | - | 3 - 4 | 270 | 63 | |
FQ215352.1 | - | 1 - 2 | 105 | 63 | ||
FQ215892.1 | - | 1 - 2 | 105 | 63 | ||
FQ216359.1 | - | 1 - 2 | 105 | 63 | ||
FQ216811.1 | - | 1 - 2 | 105 | 63 | ||
FQ217315.1 | - | 1 - 2 | 105 | 63 | ||
FQ217751.1 | - | 3 - 4 | 331 | 63 | ||
FQ219007.1 | - | 1 - 2 | 105 | 63 | ||
FQ219108.1 | - | 1 - 2 | 105 | 63 | ||
FQ219381.1 | - | 3 - 4 | 332 | 63 | ||
FQ220447.1 | - | 1 - 2 | 105 | 63 | ||
FQ220536.1 | - | 1 - 2 | 105 | 63 | ||
FQ224969.1 | - | 1 - 2 | 105 | 63 | ||
FQ227071.1 | - | 1 - 2 | 105 | 63 | ||
FQ233866.1 | - | 1 - 2 | 105 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
