Assay Details
Gene Symbol: Sar1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009622.1 | NP_001009622.1 | 5 - 6 | 426 | 76 | |
GenBank mRNA | BC088842.1 | - | 5 - 6 | 426 | 7303357 | 76 |
FQ215352.1 | - | 3 - 4 | 261 | 76 | ||
FQ215892.1 | - | 3 - 4 | 261 | 76 | ||
FQ216359.1 | - | 3 - 4 | 261 | 76 | ||
FQ216811.1 | - | 3 - 4 | 261 | 76 | ||
FQ217315.1 | - | 3 - 4 | 261 | 76 | ||
FQ217751.1 | - | 5 - 6 | 487 | 76 | ||
FQ219007.1 | - | 3 - 4 | 261 | 76 | ||
FQ219108.1 | - | 3 - 4 | 261 | 76 | ||
FQ219381.1 | - | 5 - 6 | 488 | 76 | ||
FQ220447.1 | - | 3 - 4 | 261 | 76 | ||
FQ224969.1 | - | 3 - 4 | 261 | 76 | ||
FQ227071.1 | - | 3 - 4 | 261 | 76 | ||
FQ233866.1 | - | 3 - 4 | 261 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
