Assay Details
Gene Symbol: Sec14l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109090.1 | NP_001102560.1 | 10 - 11 | 977 | 118 | |
XM_008770339.1 | XP_008768561.1 | 10 - 11 | 989 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
