Assay Details
Gene Symbol: Rnf215 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107234.1 | NP_001100704.1 | 1 - 2 | 373 | 72 | |
XM_006251300.1 | XP_006251362.1 | 1 - 2 | 399 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
