Assay Details
Gene Symbol: Ralyl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006232123.2 | XP_006232185.1 | 4 - 5 | 565 | 102 | |
XM_006232126.2 | XP_006232188.1 | 4 - 5 | 1062 | 102 | ||
XM_008760852.1 | XP_008759074.1 | 4 - 5 | 565 | 102 | ||
XM_008760853.1 | XP_008759075.1 | 4 - 5 | 1062 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
