Assay Details
Gene Symbol: Nudt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181363.2 | NP_852028.1 | 1 - 2 | 262 | 119 | |
GenBank mRNA | U58289.1 | - | 1 - 2 | 262 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
