Assay Details
Gene Symbol: Sept6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173429.1 | NP_001166900.1 | 9 - 10 | 1336 | 77 | |
XM_008773443.1 | XP_008771665.1 | 9 - 10 | 1659 | 77 | ||
GenBank mRNA | BC169051.1 | - | 9 - 10 | 1336 | 7381285 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
