Assay Details
Gene Symbol: Fdps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031840.1 | NP_114028.1 | 2 - 3 | 298 | 71 | |
GenBank mRNA | BC059125.1 | - | 2 - 3 | 226 | 71 | |
FQ209697.1 | - | 2 - 3 | 191 | 71 | ||
FQ210400.1 | - | 2 - 3 | 190 | 71 | ||
FQ211073.1 | - | 3 - 4 | 407 | 71 | ||
FQ213475.1 | - | 2 - 3 | 222 | 71 | ||
FQ218933.1 | - | 2 - 3 | 192 | 71 | ||
FQ219092.1 | - | 2 - 3 | 192 | 71 | ||
FQ225110.1 | - | 2 - 3 | 260 | 71 | ||
M17300.1 | - | 1 - 2 | 168 | 71 | ||
M34477.1 | - | 2 - 3 | 298 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
