Assay Details
Gene Symbol: Fdps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031840.1 | NP_114028.1 | 5 - 6 | 811 | 56 | |
GenBank mRNA | BC059125.1 | - | 5 - 6 | 739 | 56 | |
FQ209697.1 | - | 5 - 6 | 704 | 56 | ||
FQ210400.1 | - | 5 - 6 | 703 | 56 | ||
FQ213475.1 | - | 5 - 6 | 735 | 56 | ||
FQ218913.1 | - | 5 - 6 | 705 | 56 | ||
FQ218933.1 | - | 5 - 6 | 705 | 56 | ||
FQ219092.1 | - | 5 - 6 | 705 | 56 | ||
FQ225110.1 | - | 5 - 6 | 773 | 56 | ||
M17300.1 | - | 4 - 5 | 681 | 56 | ||
M34477.1 | - | 5 - 6 | 811 | 56 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
