Assay Details
Gene Symbol: Rad51ap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079711.1 | NP_001073179.1 | 1 - 2 | 200 | 132 | |
XM_006237502.2 | XP_006237564.1 | 1 - 2 | 180 | 132 | ||
XM_006237504.2 | XP_006237566.1 | 1 - 2 | 194 | 132 | ||
GenBank mRNA | BC128747.1 | - | 1 - 2 | 200 | 132 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC128747.1, NM_001079711.1, XM_006237502.2, XM_006237504.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
