Assay Details
Gene Symbol: Rad51ap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079711.1 | NP_001073179.1 | 2 - 3 | 259 | 161 | |
XM_006237502.2 | XP_006237564.1 | 2 - 3 | 239 | 161 | ||
XM_006237504.2 | XP_006237566.1 | 2 - 3 | 253 | 161 | ||
GenBank mRNA | BC128747.1 | - | 2 - 3 | 259 | 7099220 | 161 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
