Assay Details
Gene Symbol: Gapdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017008.4 | NP_058704.1 | 1 - 2 | 54 | 74 | |
GenBank mRNA | BC059110.1 | - | 1 - 2 | 55 | 6919611 | 74 |
X02231.1 | - | 1 - 2 | 50 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC059110.1,NM_017008.4,X02231.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
