Assay Details
Gene Symbol: Gapdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017008.4 | NP_058704.1 | 2 - 3 | 95 | 90 | |
GenBank mRNA | AB017801.1 | - | 2 - 3 | 61 | 90 | |
BC059110.1 | - | 2 - 3 | 96 | 6919611 | 90 | |
BC087743.1 | - | 2 - 3 | 64 | 7321006 | 90 | |
X02231.1 | - | 2 - 3 | 91 | 90 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AB017801.1,BC059110.1,BC087743.1,NM_017008.4,X02231.1 transcripts.
This assay is in the 5'UTR region of the AB017801.1,BC059110.1,BC087743.1,NM_017008.4,X02231.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
