Assay Details
Gene Symbol: Slco1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053441.1 | NP_445893.1 | 7 - 8 | 1152 | 146 | |
XM_006237602.1 | XP_006237664.1 | 8 - 9 | 1300 | 146 | ||
GenBank mRNA | AF306546.1 | - | 7 - 8 | 1152 | 146 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
