Assay Details
Gene Symbol: Cxcl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033882.1 | NP_001029054.1 | 1 - 2 | 145 | 156 | |
NM_001033883.1 | NP_001029055.1 | 1 - 2 | 145 | 156 | ||
NM_022177.3 | NP_071513.1 | 1 - 2 | 145 | 156 | ||
GenBank mRNA | AF209976.1 | - | 1 - 2 | 96 | 156 | |
AF217564.1 | - | 1 - 2 | 117 | 156 | ||
BC078737.1 | - | 1 - 2 | 144 | 7128141 | 156 | |
CK475409.1 | - | 1 - 2 | 145 | 156 | ||
CV081127.1 | - | 1 - 2 | 112 | 156 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF209976.1,AF217564.1,BC078737.1,NM_001033882.1,NM_001033883.1,NM_022177.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
