Assay Details
Gene Symbol: Atp6v1e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198745.2 | NP_942040.1 | 1 - 2 | 144 | 90 | |
GenBank mRNA | BC059155.1 | - | 1 - 2 | 148 | 90 | |
FQ212928.1 | - | 1 - 2 | 144 | 90 | ||
FQ213001.1 | - | 1 - 2 | 144 | 90 | ||
FQ213103.1 | - | 1 - 2 | 141 | 90 | ||
FQ213345.1 | - | 1 - 2 | 146 | 90 | ||
FQ213479.1 | - | 1 - 2 | 145 | 90 | ||
FQ213531.1 | - | 1 - 2 | 146 | 90 | ||
FQ213667.1 | - | 1 - 2 | 162 | 90 | ||
FQ213669.1 | - | 1 - 2 | 146 | 90 | ||
FQ213903.1 | - | 1 - 2 | 140 | 90 | ||
FQ214240.1 | - | 1 - 2 | 140 | 90 | ||
FQ214254.1 | - | 1 - 2 | 146 | 90 | ||
FQ214381.1 | - | 1 - 2 | 144 | 90 | ||
FQ214406.1 | - | 1 - 2 | 139 | 90 | ||
FQ214440.1 | - | 1 - 2 | 146 | 90 | ||
FQ214476.1 | - | 1 - 2 | 149 | 90 | ||
FQ214497.1 | - | 1 - 2 | 112 | 90 | ||
FQ214920.1 | - | 1 - 2 | 144 | 90 | ||
FQ216629.1 | - | 1 - 2 | 144 | 90 | ||
FQ224865.1 | - | 1 - 2 | 145 | 90 | ||
FQ226034.1 | - | 1 - 2 | 139 | 90 | ||
FQ230006.1 | - | 1 - 2 | 144 | 90 | ||
FQ230733.1 | - | 1 - 2 | 139 | 90 | ||
FQ233569.1 | - | 1 - 2 | 146 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
