Assay Details
Gene Symbol: Atp6v1e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198745.2 | NP_942040.1 | 2 - 3 | 201 | 129 | |
GenBank mRNA | BC059155.1 | - | 2 - 3 | 205 | 129 | |
FQ212928.1 | - | 2 - 3 | 201 | 129 | ||
FQ213001.1 | - | 2 - 3 | 201 | 129 | ||
FQ213103.1 | - | 2 - 3 | 198 | 129 | ||
FQ213345.1 | - | 2 - 3 | 203 | 129 | ||
FQ213479.1 | - | 2 - 3 | 202 | 129 | ||
FQ213531.1 | - | 2 - 3 | 203 | 129 | ||
FQ213667.1 | - | 2 - 3 | 219 | 129 | ||
FQ213669.1 | - | 2 - 3 | 203 | 129 | ||
FQ213903.1 | - | 2 - 3 | 197 | 129 | ||
FQ214240.1 | - | 2 - 3 | 197 | 129 | ||
FQ214254.1 | - | 2 - 3 | 203 | 129 | ||
FQ214381.1 | - | 2 - 3 | 201 | 129 | ||
FQ214406.1 | - | 2 - 3 | 196 | 129 | ||
FQ214440.1 | - | 2 - 3 | 203 | 129 | ||
FQ214476.1 | - | 2 - 3 | 206 | 129 | ||
FQ214497.1 | - | 2 - 3 | 169 | 129 | ||
FQ214920.1 | - | 2 - 3 | 201 | 129 | ||
FQ216629.1 | - | 2 - 3 | 201 | 129 | ||
FQ226034.1 | - | 2 - 3 | 196 | 129 | ||
FQ230006.1 | - | 2 - 3 | 201 | 129 | ||
FQ230733.1 | - | 2 - 3 | 196 | 129 | ||
FQ233079.1 | - | 2 - 3 | 201 | 129 | ||
FQ233569.1 | - | 2 - 3 | 203 | 129 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
