Assay Details
Gene Symbol: Ndufa9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100752.1 | NP_001094222.1 | 1 - 2 | 68 | 97 | |
GenBank mRNA | BC168777.1 | - | 1 - 2 | 68 | 97 | |
FQ216233.1 | - | 1 - 2 | 68 | 97 | ||
FQ216540.1 | - | 1 - 2 | 68 | 97 | ||
FQ216671.1 | - | 1 - 2 | 58 | 97 | ||
FQ216764.1 | - | 1 - 2 | 58 | 97 | ||
FQ232722.1 | - | 1 - 2 | 66 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
