Assay Details
Gene Symbol: Ndufa9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100752.1 | NP_001094222.1 | 7 - 8 | 731 | 122 | |
XM_006237500.1 | XP_006237562.1 | 7 - 8 | 814 | 122 | ||
GenBank mRNA | BC091192.1 | - | 7 - 8 | 697 | 122 | |
BC168777.1 | - | 7 - 8 | 731 | 6922152 | 122 | |
FQ216233.1 | - | 7 - 8 | 731 | 122 | ||
FQ216540.1 | - | 7 - 8 | 731 | 123 | ||
FQ216671.1 | - | 7 - 8 | 721 | 122 | ||
FQ216764.1 | - | 7 - 8 | 721 | 122 | ||
FQ232722.1 | - | 7 - 8 | 729 | 122 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
