Assay Details
Gene Symbol: Olr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133306.2 | NP_579840.2 | 4 - 5 | 648 | 89 | |
GenBank mRNA | AB005900.1 | - | 4 - 5 | 648 | 89 | |
BC097290.1 | - | 4 - 5 | 601 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
