Assay Details
Gene Symbol: Bicd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108653.1 | NP_001102123.1 | 3 - 4 | 1009 | 115 | |
XM_006237703.2 | XP_006237765.1 | 3 - 4 | 989 | 115 | ||
XM_006237705.2 | XP_006237767.1 | 3 - 4 | 989 | 115 | ||
XM_006237706.2 | XP_006237768.1 | 3 - 4 | 989 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
