Assay Details
Gene Symbol: Amn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008333.2 | NP_001008334.2 | 1 - 2 | 90 | 135 | |
GenBank mRNA | BC086357.1 | - | 1 - 2 | 60 | 135 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC086357.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
