Assay Details
Gene Symbol: Amn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008333.2 | NP_001008334.2 | 4 - 5 | 579 | 65 | |
XM_006237700.1 | XP_006237762.1 | 4 - 5 | 1065 | 65 | ||
GenBank mRNA | BC086357.1 | - | 4 - 5 | 549 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
