Assay Details
Gene Symbol: Amn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008333.2 | NP_001008334.2 | 6 - 7 | 749 | 156 | |
XM_006237700.1 | XP_006237762.1 | 6 - 7 | 1235 | 156 | ||
GenBank mRNA | BC086357.1 | - | 6 - 7 | 719 | 156 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P382363 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541341 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452293 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180124 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103367 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345934 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393622 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 970776 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691792 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951978 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832833 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357248 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550937 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 63985 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385524 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5004 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P382363 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P382363 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P382363 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P382363 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 22811 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 59123 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 96945 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10867016 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867014 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867017 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867013 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867015 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867012 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867011 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10867010 | - | - |
Rat Genome 230 2.0 Array | 1380192_PM_at | - | - |
Rat Genome 230 2.0 Array | 1381961_at | - | - |
Rat Genome 230 2.0 Array | 1376714_PM_at | - | - |
Rat Genome 230 2.0 Array | 1381961_PM_at | - | - |
Rat Genome 230 2.0 Array | 1380192_at | - | - |
Rat Genome 230 2.0 Array | 1376714_at | - | - |
Affymetrix HT Rat Focus Array | 1376714_at | - | - |
Affymetrix HT Rat Focus Array | 1376714_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1380192_at | - | - |
Affymetrix HT Rat Focus Array | 1380192_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0400029393.rn.1 | TC0400004138.rn.1 | RTA TC |
Rat Expression Set 230 | 1381961_PM_at | - | - |
Rat Expression Set 230 | 1380192_at | - | - |
Rat Expression Set 230 | 1381961_at | - | - |
Rat Expression Set 230 | 1376714_PM_at | - | - |
Rat Expression Set 230 | 1376714_at | - | - |
Rat Expression Set 230 | 1380192_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867010 | - | - |
Rat Genome U34 Set | rc_AI236072_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21811580 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357643 | - | - |
RatRef-12 Expression BeadChip | ILMN_70160 | - | - |
