Assay Details
Gene Symbol: Eno2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139325.3 | NP_647541.1 | 10 - 11 | 1247 | 71 | |
XM_006237330.2 | XP_006237392.1 | 10 - 11 | 1533 | 71 | ||
GenBank mRNA | AF019973.1 | - | 10 - 11 | 1247 | 71 | |
BC060310.1 | - | 10 - 11 | 1238 | 6888358 | 71 | |
FQ214264.1 | - | 10 - 11 | 1222 | 71 | ||
M11931.1 | - | 10 - 11 | 1247 | 71 |
Gene Symbol: LOC100911625 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750657.3 | XP_003750705.1 | 10 - 11 | 1303 | 71 | |
XM_006244636.2 | XP_006244698.1 | 10 - 11 | 1535 | 71 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
