Assay Details
Gene Symbol: Phc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107886.1 | NP_001101356.1 | 2 - 3 | 242 | 113 | |
XM_006237300.2 | XP_006237362.1 | 3 - 4 | 267 | 113 | ||
XM_006237302.2 | XP_006237364.1 | 1 - 2 | 165 | 113 | ||
XM_006237303.2 | XP_006237365.1 | 3 - 4 | 263 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
