Assay Details
Gene Symbol: Phc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107886.1 | NP_001101356.1 | 6 - 7 | 741 | 88 | |
XM_006237300.2 | XP_006237362.1 | 7 - 8 | 766 | 88 | ||
XM_006237302.2 | XP_006237364.1 | 5 - 6 | 664 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
