Assay Details
Gene Symbol: Adipor2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037979.1 | NP_001033068.1 | 4 - 5 | 656 | 131 | |
XM_006237183.1 | XP_006237245.1 | 5 - 6 | 962 | 131 | ||
XM_006237184.2 | XP_006237246.1 | 5 - 6 | 833 | 131 | ||
XM_006237185.2 | XP_006237247.1 | 5 - 6 | 848 | 131 | ||
XM_006237186.2 | XP_006237248.1 | 5 - 6 | 977 | 131 | ||
GenBank mRNA | AY724514.1 | - | 5 - 6 | 916 | 131 | |
DQ148392.1 | - | 4 - 5 | 656 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
