Assay Details
Gene Symbol: Cd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053018.1 | NP_444177.1 | 2 - 3 | 236 | 94 | |
GenBank mRNA | BC161804.1 | - | 2 - 3 | 211 | 94 | |
CF111078.1 | - | 2 - 3 | 245 | 94 | ||
FQ212497.1 | - | 2 - 3 | 260 | 94 | ||
FQ213039.1 | - | 2 - 3 | 285 | 94 | ||
FQ215636.1 | - | 2 - 3 | 248 | 94 | ||
FQ216402.1 | - | 2 - 3 | 285 | 94 | ||
FQ216770.1 | - | 2 - 3 | 272 | 94 | ||
FQ219799.1 | - | 2 - 3 | 259 | 94 | ||
FQ220476.1 | - | 2 - 3 | 260 | 94 | ||
FQ224453.1 | - | 2 - 3 | 260 | 94 | ||
FQ229965.1 | - | 2 - 3 | 307 | 94 | ||
FQ230141.1 | - | 2 - 3 | 322 | 94 | ||
KC162232.1 | - | 175 | 94 | |||
KC162233.1 | - | 1 - 2 | 222 | 94 | ||
X76489.1 | - | 2 - 3 | 175 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
