Assay Details
Gene Symbol: Cd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053018.1 | NP_444177.1 | 5 - 6 | 505 | 130 | |
GenBank mRNA | BC161804.1 | - | 5 - 6 | 480 | 9101765 | 130 |
BG670145.1 | - | 1 - 2 | 97 | 130 | ||
BQ202980.1 | - | 3 - 4 | 130 | |||
CF111078.1 | - | 5 - 6 | 514 | 130 | ||
FQ212497.1 | - | 5 - 6 | 529 | 130 | ||
FQ213039.1 | - | 5 - 6 | 554 | 130 | ||
FQ213302.1 | - | 5 - 6 | 523 | 130 | ||
FQ215636.1 | - | 5 - 6 | 517 | 130 | ||
FQ216402.1 | - | 5 - 6 | 554 | 130 | ||
FQ216770.1 | - | 5 - 6 | 541 | 130 | ||
FQ224453.1 | - | 5 - 6 | 529 | 130 | ||
FQ229965.1 | - | 5 - 6 | 576 | 130 | ||
FQ230141.1 | - | 5 - 6 | 591 | 130 | ||
X76489.1 | - | 5 - 6 | 444 | 130 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
