Assay Details
Gene Symbol: Rimklb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006237315.2 | XP_006237377.1 | 3 - 4 | 2628 | 78 | |
XM_006237316.2 | XP_006237378.1 | 3 - 4 | 585 | 78 | ||
XM_006237317.2 | XP_006237379.1 | 3 - 4 | 622 | 78 | ||
XM_008763276.1 | XP_008761498.1 | 4 - 5 | 810 | 78 | ||
XM_342749.7 | XP_342750.3 | 3 - 4 | 543 | 78 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
