Assay Details
Gene Symbol: Rimklb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006237315.2 | XP_006237377.1 | 4 - 5 | 2716 | 124 | |
XM_006237316.2 | XP_006237378.1 | 4 - 5 | 673 | 124 | ||
XM_006237317.2 | XP_006237379.1 | 4 - 5 | 710 | 124 | ||
XM_008763276.1 | XP_008761498.1 | 5 - 6 | 898 | 124 | ||
XM_342749.7 | XP_342750.3 | 4 - 5 | 631 | 124 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
