Assay Details
Gene Symbol: Rimklb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006237315.2 | XP_006237377.1 | 1 - 2 | 2175 | 128 | |
XM_008763276.1 | XP_008761498.1 | 2 - 3 | 357 | 128 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006237315.2, XM_008763276.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
