Assay Details
Gene Symbol: Slc6a12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017335.1 | NP_059031.1 | 11 - 12 | 1670 | 84 | |
GenBank mRNA | FQ229573.1 | - | 1194 | 84 | ||
U28927.1 | - | 11 - 12 | 1670 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
