Assay Details
Gene Symbol: Bcl2l14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024338.1 | NP_001019509.1 | 2 - 3 | 215 | 108 | |
XM_006237533.2 | XP_006237595.1 | 3 - 4 | 361 | 108 | ||
GenBank mRNA | BC079010.1 | - | 2 - 3 | 215 | 7113223 | 108 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC079010.1, NM_001024338.1, XM_006237533.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
