Assay Details
Gene Symbol: Bcl2l14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024338.1 | NP_001019509.1 | 3 - 4 | 647 | 135 | |
XM_006237533.2 | XP_006237595.1 | 4 - 5 | 793 | 135 | ||
XM_006237534.2 | XP_006237596.1 | 2 - 3 | 652 | 135 | ||
XM_008763416.1 | XP_008761638.1 | 2 - 3 | 477 | 135 | ||
GenBank mRNA | BC079010.1 | - | 3 - 4 | 647 | 7113223 | 135 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
