Assay Details
Gene Symbol: Mfap5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108644.1 | NP_001102114.1 | 9 - 10 | 455 | 83 | |
GenBank mRNA | FQ229611.1 | - | 8 - 9 | 561 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
