Assay Details
Gene Symbol: Fdxr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024153.1 | NP_077067.1 | 11 - 12 | 1376 | 130 | |
GenBank mRNA | BC088844.1 | - | 11 - 12 | 1383 | 130 | |
D63761.1 | - | 11 - 12 | 1376 | 130 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
