Assay Details
Gene Symbol: Vmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138839.2 | NP_620194.1 | 4 - 5 | 414 | 102 | |
XM_006247078.2 | XP_006247140.1 | 4 - 5 | 892 | 102 | ||
XM_006247079.2 | XP_006247141.1 | 5 - 6 | 436 | 102 | ||
GenBank mRNA | AF411216.1 | - | 4 - 5 | 360 | 102 | |
BC061721.1 | - | 4 - 5 | 414 | 102 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
