Assay Details
Gene Symbol: Shbg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008767771.1 | XP_008765993.1 | 2 | 296 | 96 | |
XM_008767772.1 | XP_008765994.1 | 1 | 104 | 96 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
