Assay Details
Gene Symbol: Psmd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008281.1 | NP_001008282.1 | 2 - 3 | 544 | 72 | |
XM_006247272.1 | XP_006247334.1 | 2 - 3 | 759 | 72 | ||
GenBank mRNA | BC085881.1 | - | 2 - 3 | 544 | 7191926 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
