Assay Details
Gene Symbol: Nubp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011891.1 | NP_001011891.1 | 1 - 2 | 41 | 92 | |
XM_008767558.1 | XP_008765780.1 | 1 - 2 | 37 | 92 | ||
GenBank mRNA | BC079386.1 | - | 1 - 2 | 41 | 7111553 | 92 |
FQ213066.1 | - | 1 - 2 | 55 | 92 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC079386.1,NM_001011891.1,XM_008767558.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
